Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-N,3-dimethyl-2-pyridinamine ≥97% research grade

Supplier: AOB CHEM USA
Warning

17963-1G 17963-25G 17963-500MG 17963-5G
76883-866EA 138.98 USD InStock
76883-866 76883-868 76883-870 76883-872
5-Bromo-N,3-dimethyl-2-pyridinamine ≥97% research grade
5-Bromo-N,3-dimethyl-2-pyridinamine


Formula: C₇H₉BrN₂
MW: 201.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10000482
CAS Number: 245765-66-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty