Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,6-difluoro-4-methylphenyl)boronic acid 95% research grade

Supplier: AOB CHEM USA
Warning

18706-1G 18706-500MG 18706-5G 18706-25G
76884-122EA 500.75 USD InStock
76884-122 76884-124 76884-126 77657-808
(2,6-difluoro-4-methylphenyl)boronic acid 95% research grade
(2,6-difluoro-4-methylphenyl)boronic acid


Formula: C₇H₇BF₂O₂
MW: 171.937 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072611
CAS Number: 2246867-26-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty