Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,4-Dichloro-6-methylphenyl)boronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

20261-1G 20261-500MG 20261-5G 20261-25G
76884-480EA 553.79 USD InStock
76884-480 76884-482 76884-484 77654-100
(2,4-Dichloro-6-methylphenyl)boronic acid ≥95% research grade
(2,4-Dichloro-6-methylphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₇H₇BCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28787026
CAS Number: 3028870-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty