Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,5-dichloro-4-methylbenzene 97% research grade

Supplier: AOB CHEM USA
Warning

21151-1G 21151-500MG 21151-5G 21151-25G
76884-736EA 471.04 USD InStock
76884-736 76884-738 76884-740 77654-162
1-Bromo-2,5-dichloro-4-methylbenzene 97% research grade
1-Bromo-2,5-dichloro-4-methylbenzene
Precaución: for Research Use Only


Formula: C₇H₅BrCl₂
Storage Temperature: Ambient
MDL Number: MFCD26131442
CAS Number: 1807208-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty