Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Chloro-2-(methoxymethoxy)phenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

21643-1G 21643-25G 21643-500MG 21643-5G
76884-820EA 203.7 USD InStock
76884-820 76884-822 76884-824 76884-826
(4-Chloro-2-(methoxymethoxy)phenyl)boronic acid ≥97% research grade
(4-Chloro-2-(methoxymethoxy)phenyl)boronic acid


Formula: C₈H₁₀BClO₄
MW: 216.427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34182896
CAS Number: 2275572-12-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty