Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(ethenyloxy)benzene ≥97%

Supplier: AOB CHEM USA
Warning

22035-1G 22035-25G 22035-500MG 22035-5G
76884-912EA 173.99 USD InStock
76884-912 76884-914 76884-916 76884-918
1-Bromo-4-(ethenyloxy)benzene ≥97%
1-Bromo-4-(ethenyloxy)benzene
Precaución: for Research Use Only


Formula: C₈H₇BrO
Storage Temperature: Refrigerator
MDL Number: MFCD21755355
CAS Number: 1005-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty