Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylthio)thiophene-3-carboxylic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

23715-1G 23715-25G 23715-500MG 23715-5G
76885-306EA 203.7 USD InStock
76885-306 76885-308 76885-310 76885-312
2-(Methylthio)thiophene-3-carboxylic acid ≥97% research grade
2-(Methylthio)thiophene-3-carboxylic acid


Formula: C₆H₆O₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD01859894
CAS Number: 71154-30-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty