Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dichloro-5-(3-nitrophenyl)benzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

23851-1G 23851-25G 23851-5G 23851-500MG
76885-344EA 109.28 USD InStock
76885-344 76885-346 76885-348 77645-384
1,3-Dichloro-5-(3-nitrophenyl)benzene ≥97% research grade
1,3-Dichloro-5-(3-nitrophenyl)benzene


Formula: C₁₂H₇Cl₂NO₂
MW: 268.095 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21332977
CAS Number: 859940-64-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty