Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-Fluoro-5-(Methylsulfanyl)Benzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

24556-1G 24556-25G 24556-500MG 24556-5G
76885-490EA 119.89 USD InStock
76885-490 76885-492 76885-494 76885-496
1-Bromo-3-Fluoro-5-(Methylsulfanyl)Benzene ≥97% research grade
1-Bromo-3-Fluoro-5-(Methylsulfanyl)Benzene


Formula: C₇H₆BrFS
Storage Temperature: Ambient
MDL Number: MFCD11848446
CAS Number: 453566-03-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty