Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethoxypyridine-4-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

24732-1G 24732-25G 24732-500MG 24732-5G
76885-538EA 124.13 USD InStock
76885-538 76885-540 76885-542 76885-544
2,3-Dimethoxypyridine-4-carboxylic acid ≥97%
2,3-Dimethoxypyridine-4-carboxylic acid


Formula: C₈H₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD10697544
CAS Number: 944906-83-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty