Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromobenzyl)-diethylamine ≥97% research grade

Supplier: AOB CHEM USA
Warning

25329-1G 25329-25G 25329-5G 25329-500MG
76885-636EA 88.06 USD InStock
76885-636 76885-638 76885-640 77645-470
(3-Bromobenzyl)-diethylamine ≥97% research grade
(3-Bromobenzyl)-diethylamine


Formula: C₁₁H₁₆BrN
MW: 242.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08276025
CAS Number: 27958-94-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty