Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,6-Dichloropyridin-4-yl)(morpholino)methanone ≥97% research grade

Supplier: AOB CHEM USA
Warning

26000-1G 26000-25G 26000-500MG 26000-5G
76885-858EA 367.08 USD InStock
76885-858 76885-860 76885-862 76885-864
(2,6-Dichloropyridin-4-yl)(morpholino)methanone ≥97% research grade
(2,6-Dichloropyridin-4-yl)(morpholino)methanone


Formula: C₁₀H₁₀Cl₂N₂O₂
MW: 261.105 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01714001
CAS Number: 57803-44-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty