Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromo-2-methoxy-6-fluorophenyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

26045-1G 26045-25G 26045-500MG 26045-5G
76885-888EA 185.66 USD InStock
76885-888 76885-890 76885-892 76885-894
(4-Bromo-2-methoxy-6-fluorophenyl)methanol ≥97%
(4-Bromo-2-methoxy-6-fluorophenyl)methanol


Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD24848696
CAS Number: 1417736-85-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty