Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-methyl-4-phenylbenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

26499-1G 26499-25G 26499-500MG 26499-5G
76886-006EA 237.65 USD InStock
76886-006 76886-008 76886-010 76886-012
1-Bromo-2-methyl-4-phenylbenzene ≥97% research grade
1-Bromo-2-methyl-4-phenylbenzene


Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00089406
CAS Number: 92022-07-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty