Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(5-butylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOB CHEM USA
Warning

28327-25G 28327-5G
76886-410EA 453.01 USD InStock
76886-410 76886-412
2-(5-butylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(5-butylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane


Formula: C₁₄H₂₃BO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD28154573
CAS Number: 1338083-26-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty