Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(5-Bromo-2-fluorophenyl)-1H-imidazole ≥97% research grade

Supplier: AOB CHEM USA
Warning

28521-1G 28521-25G 28521-500MG 28521-5G
76886-470EA 361.77 USD InStock
76886-470 76886-472 76886-474 76886-476
1-(5-Bromo-2-fluorophenyl)-1H-imidazole ≥97% research grade
1-(5-Bromo-2-fluorophenyl)-1H-imidazole


Formula: C₉H₆BrFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD20919805
CAS Number: 425379-84-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty