Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methoxymethyl)-thiophene-5-boronic acid pinacol ester ≥95% research grade

Supplier: AOB CHEM USA
Warning

28784-1G 28784-25G 28784-500MG 28784-5G
76886-556EA 320.4 USD InStock
76886-556 76886-558 76886-560 76886-562
2-(Methoxymethyl)-thiophene-5-boronic acid pinacol ester ≥95% research grade
2-(Methoxymethyl)-thiophene-5-boronic acid pinacol ester


Formula: C₁₂H₁₉BO₃S
MW: 254.153 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28388512
CAS Number: 2102681-58-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty