Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-isopropoxy-4-(trifluoromethyl)benzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

29206-1G 29206-25G 29206-500MG 29206-5G
76886-684EA 193.09 USD InStock
76886-684 76886-686 76886-688 76886-690
1-Bromo-2-isopropoxy-4-(trifluoromethyl)benzene ≥97% research grade
1-Bromo-2-isopropoxy-4-(trifluoromethyl)benzene


Formula: C₁₀H₁₀BrF₃O
MW: 283.085 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20441887
CAS Number: 1345471-36-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty