Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,5-difluoroiodobenzene ≥97%

Supplier: AOB CHEM USA
Warning

29475-1G 29475-25G 29475-500MG 29475-5G
76886-742EA 200.52 USD InStock
76886-742 76886-744 76886-746 76886-748
2-Bromo-4,5-difluoroiodobenzene ≥97%
2-Bromo-4,5-difluoroiodobenzene


Formula: C₆H₂BrF₂I
Storage Temperature: Refrigerator
MDL Number: MFCD20233325
CAS Number: 1349718-22-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty