Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',4'-dichloro-[1,1'-biphenyl]-4-ol ≥97%

Supplier: AOB CHEM USA
Warning

29558-1G 29558-25G 29558-500MG 29558-5G
76886-758EA 126.25 USD InStock
76886-758 76886-760 76886-762 76886-764
2',4'-dichloro-[1,1'-biphenyl]-4-ol ≥97%
2',4'-dichloro-[1,1'-biphenyl]-4-ol


Formula: C₁₂H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD11574823
CAS Number: 53890-76-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty