Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chloro-3-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: AOB CHEM USA
Warning

29658-1G 29658-25G 29658-500MG 29658-5G
76886-788EA 432.85 USD InStock
76886-788 76886-790 76886-792 76886-794
2-(4-Chloro-3-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(4-Chloro-3-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane


Formula: C₁₅H₂₂BClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18732294
CAS Number: 2121511-69-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty