Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1-Methyl-1H-pyrazol-5-yl)ethanone 95% research grade

Supplier: AOB CHEM USA
Warning

30702-1G 30702-25G 30702-5G 30702-500MG
76886-966EA 102.91 USD InStock
76886-966 76886-968 76886-970 77645-900
1-(1-Methyl-1H-pyrazol-5-yl)ethanone 95% research grade
1-(1-Methyl-1H-pyrazol-5-yl)ethanone


Formula: C₆H₈N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD04968784
CAS Number: 137890-05-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty