Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-5-methylanisole ≥97%

Supplier: AOB CHEM USA
Warning

31458-1G 31458-25G 31458-500MG 31458-5G
76887-270EA 198.39 USD InStock
76887-270 76887-272 76887-274 76887-276
2-Iodo-5-methylanisole ≥97%
2-Iodo-5-methylanisole
Precaución: for Research Use Only


Formula: C₈H₉IO
MW: 248.0609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547792
CAS Number: 186583-59-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty