Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-2-(2-(methylsulfonyl)phenyl)-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOB CHEM USA
Warning

31616-1G 31616-25G 31616-500MG 31616-5G
76887-308EA 264.17 USD InStock
76887-308 76887-310 76887-312 76887-314
4,4,5,5-Tetramethyl-2-(2-(methylsulfonyl)phenyl)-1,3,2-dioxaborolane ≥97% research grade
4,4,5,5-Tetramethyl-2-(2-(methylsulfonyl)phenyl)-1,3,2-dioxaborolane


Formula: C₁₃H₁₉BO₄S
MW: 282.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14584689
CAS Number: 879648-22-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty