Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-2-iodobenzyl alcohol ≥97%

Supplier: AOB CHEM USA
Warning

31986-1G 31986-25G 31986-500MG 31986-5G
76887-354EA 177.18 USD InStock
76887-354 76887-356 76887-358 76887-360
3-Fluoro-2-iodobenzyl alcohol ≥97%
3-Fluoro-2-iodobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD18396946
CAS Number: 1261827-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty