Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Chloro-2-(hydroxymethyl)phenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

32866-1G 32866-25G 32866-500MG 32866-5G
76887-498EA 291.75 USD InStock
76887-498 76887-500 76887-502 76887-504
(4-Chloro-2-(hydroxymethyl)phenyl)boronic acid ≥97% research grade
(4-Chloro-2-(hydroxymethyl)phenyl)boronic acid


Formula: C₇H₈BClO₃
MW: 186.401 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23163242
CAS Number: 1365244-07-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty