Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Isopropoxypyridin-4-yl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

33239-1G 33239-25G 33239-500MG 33239-5G
76887-556EA 218.55 USD InStock
76887-556 76887-558 76887-560 76887-562
(2-Isopropoxypyridin-4-yl)methanol ≥97%
(2-Isopropoxypyridin-4-yl)methanol


Formula: C₉H₁₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD14606470
CAS Number: 754218-88-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty