Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-6-hydroxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

34382-1G 34382-25G 34382-500MG 34382-5G
76887-848EA 155.96 USD InStock
76887-848 76887-850 76887-852 76887-854
2,3-Dichloro-6-hydroxybenzoic acid ≥97% research grade
2,3-Dichloro-6-hydroxybenzoic acid


Formula: C₇H₄Cl₂O₃
MW: 207.011 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28786450
CAS Number: 1806282-06-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty