Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-4-methoxybenzyl alcohol ≥95%

Supplier: AOB CHEM USA
Warning

34605-1G 34605-25G 34605-5G
76887-872EA 62.6 USD InStock
76887-872 76887-874 76887-876
2,3-Difluoro-4-methoxybenzyl alcohol ≥95%
2,3-Difluoro-4-methoxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115984
CAS Number: 886500-71-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty