Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-N-methylbenzamide ≥97%

Supplier: AOB CHEM USA
Warning

36035-1G 36035-25G 36035-500MG 36035-5G
76888-182EA 120.95 USD InStock
76888-182 76888-184 76888-186 76888-188
2,3-Difluoro-N-methylbenzamide ≥97%
2,3-Difluoro-N-methylbenzamide


Formula: C₈H₇F₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD16836083
CAS Number: 1250397-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty