Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Bromo-6-chloropyridin-2-yl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

36382-1G 36382-25G 36382-500MG 36382-5G
76888-384EA 213.25 USD InStock
76888-384 76888-386 76888-388 76888-390
(5-Bromo-6-chloropyridin-2-yl)methanol ≥97%
(5-Bromo-6-chloropyridin-2-yl)methanol


Formula: C₆H₅BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD16606987
CAS Number: 1227563-64-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty