Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-4,5-dimethoxybenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

36931-1G 36931-500MG 36931-5G
76888-746EA 689.59 USD InStock
76888-746 76888-748 76888-750
2,3-Difluoro-4,5-dimethoxybenzaldehyde ≥97%
2,3-Difluoro-4,5-dimethoxybenzaldehyde


Formula: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28365519
CAS Number: 172657-04-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty