Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Difluoromethoxy)-2-fluorobenzene ≥97%

Supplier: AOB CHEM USA
Warning

36987-1G 36987-25G 36987-500MG 36987-5G
76888-778EA 379.81 USD InStock
76888-778 76888-780 76888-782 76888-784
1-(Difluoromethoxy)-2-fluorobenzene ≥97%
1-(Difluoromethoxy)-2-fluorobenzene


Formula: C₇H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD13194314
CAS Number: 97963-49-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty