Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Fluoro-6-Methoxyphenyl)(Methyl)Sulfane ≥97% research grade

Supplier: AOB CHEM USA
Warning

37191-1G 37191-25G 37191-500MG 37191-5G
76888-874EA 171.87 USD InStock
76888-874 76888-876 76888-878 76888-880
(2-Fluoro-6-Methoxyphenyl)(Methyl)Sulfane ≥97% research grade
(2-Fluoro-6-Methoxyphenyl)(Methyl)Sulfane


Formula: C₈H₉FOS
Storage Temperature: Refrigerator
MDL Number: MFCD18904713
CAS Number: 1378573-53-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty