Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-3-(3-chloro-4-methoxyphenyl)acrylic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

37241-1G 37241-25G 37241-5G
76888-906EA 62.6 USD InStock
76888-906 76888-908 76888-910
(E)-3-(3-chloro-4-methoxyphenyl)acrylic acid ≥97% research grade
(E)-3-(3-chloro-4-methoxyphenyl)acrylic acid


Formula: C₁₀H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD05739914
CAS Number: 58236-76-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty