Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(trifluoromethyl)cinnamic acid ≥97%

Supplier: AOB CHEM USA
Warning

37244-1G 37244-25G 37244-5G
76888-918EA 73.21 USD InStock
76888-918 76888-920 76888-922
4-Chloro-2-(trifluoromethyl)cinnamic acid ≥97%
4-Chloro-2-(trifluoromethyl)cinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₆ClF₃O₂
MW: 250.602 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06205746
CAS Number: 773132-29-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty