Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-5-methoxy-4-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

37732-1G 37732-500MG 37732-5G
76889-144EA 587.74 USD InStock
76889-144 76889-146 76889-148
2,3-Difluoro-5-methoxy-4-methylbenzoic acid ≥97%
2,3-Difluoro-5-methoxy-4-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₈F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD27986839
CAS Number: 1706446-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty