Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(p-Tolyl)imidazo[1,2-a]pyridine ≥97%

Supplier: AOB CHEM USA
Warning

37974-1G 37974-25G 37974-500MG 37974-5G
76889-250EA 350.1 USD InStock
76889-250 76889-252 76889-254 76889-256
2-(p-Tolyl)imidazo[1,2-a]pyridine ≥97%
2-(p-Tolyl)imidazo[1,2-a]pyridine


Formula: C₁₄H₁₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD00452508
CAS Number: 65964-60-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty