Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(Trifluoromethyl)phenyl)-1,3,4-oxadiazole 97% research grade

Supplier: AOB CHEM USA
Warning

38383-1G 38383-25G 38383-5G 38383-500MG
76889-522EA 94.43 USD InStock
76889-522 76889-524 76889-526 77646-560
2-(4-(Trifluoromethyl)phenyl)-1,3,4-oxadiazole 97% research grade
2-(4-(Trifluoromethyl)phenyl)-1,3,4-oxadiazole


Formula: C₉H₅F₃N₂O
MW: 214.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01814630
CAS Number: 477886-85-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty