Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-fluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

40212-1G 40212-25G 40212-500MG 40212-5G
76889-782EA 380.87 USD InStock
76889-782 76889-784 76889-786 76889-788
2,4-Dichloro-5-fluorophenylboronic acid ≥97%
2,4-Dichloro-5-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₄BCl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD19981520
CAS Number: 1417914-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty