Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

40528-25G 40528-5G
76889-796EA 215.37 USD InStock
76889-796 76889-798
4-Ethylphenylboronic acid pinacol ester ≥97%
4-Ethylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₁BO₂
Storage Temperature: Ambient
MDL Number: MFCD18643353
CAS Number: 1075719-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty