Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,3-Dichloro-4-ethoxyphenyl)boronic acid 97%

Supplier: AOB CHEM USA
Warning

40921-1G 40921-25G 40921-5G 40921-500MG
76889-822EA 102.91 USD InStock
76889-822 76889-824 76889-826 77650-594
(2,3-Dichloro-4-ethoxyphenyl)boronic acid 97%
(2,3-Dichloro-4-ethoxyphenyl)boronic acid


Formula: C₈H₉BCl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD21609523
CAS Number: 2096341-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty