Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Hydroxymethyl)-4-(trifluoromethyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

41006-1G 41006-25G 41006-500MG 41006-5G
76889-836EA 242.95 USD InStock
76889-836 76889-838 76889-840 76889-842
2-(Hydroxymethyl)-4-(trifluoromethyl)phenylboronic acid ≥97% research grade
2-(Hydroxymethyl)-4-(trifluoromethyl)phenylboronic acid


Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22574938
CAS Number: 1629148-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty