Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chloro-4,5-dimethylphenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

45098-1G 45098-25G 45098-500MG 45098-5G
76889-916EA 375.56 USD InStock
76889-916 76889-918 76889-920 76889-922
(2-Chloro-4,5-dimethylphenyl)boronic acid ≥97% research grade
(2-Chloro-4,5-dimethylphenyl)boronic acid


Formula: C₈H₁₀BClO₂
Storage Temperature: Ambient
MDL Number: MFCD24217817
CAS Number: 2246683-46-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty