Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Fluoropyridin-4-yl)ethanone ≥97%

Supplier: AOB CHEM USA
Warning

45971-1G 45971-25G 45971-500MG 45971-5G
76890-062EA 152.77 USD InStock
76890-062 76890-064 76890-066 76890-068
1-(3-Fluoropyridin-4-yl)ethanone ≥97%
1-(3-Fluoropyridin-4-yl)ethanone


Formula: C₇H₆FNO
Storage Temperature: Refrigerator
MDL Number: MFCD07375077
CAS Number: 87674-21-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty