Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(cyclopropylmethyl)benzene ≥97%

Supplier: AOB CHEM USA
Warning

46463-1G 46463-25G 46463-500MG 46463-5G
76890-186EA 291.75 USD InStock
76890-186 76890-188 76890-190 76890-192
1-Bromo-4-(cyclopropylmethyl)benzene ≥97%
1-Bromo-4-(cyclopropylmethyl)benzene


Formula: C₁₀H₁₁Br
Storage Temperature: Refrigerator
MDL Number: MFCD12030958
CAS Number: 179251-28-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty