Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Biphenyl-3-carboxaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

46723-25G 46723-5G
76890-228EA 225.98 USD InStock
76890-228 76890-230
Biphenyl-3-carboxaldehyde ≥97% research grade
Biphenyl-3-carboxaldehyde


Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01740432
CAS Number: 1204-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty