Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(6-Bromopyridin-2-yl)acetic acid ≥97%

Supplier: AOB CHEM USA
Warning

48524-1G 48524-25G 48524-500MG 48524-5G
76890-526EA 188.85 USD InStock
76890-526 76890-528 76890-530 76890-532
2-(6-Bromopyridin-2-yl)acetic acid ≥97%
2-(6-Bromopyridin-2-yl)acetic acid
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD10000075
CAS Number: 1093879-46-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty