Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dibromo-4-chloro-5-fluorobenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

49963-1G 49963-25G 49963-500MG 49963-5G
76890-880EA 185.66 USD InStock
76890-880 76890-882 76890-884 76890-886
1,2-Dibromo-4-chloro-5-fluorobenzene ≥97% research grade
1,2-Dibromo-4-chloro-5-fluorobenzene


Formula: C₆H₂Br₂ClF
MW: 288.339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD9878131
CAS Number: 1000578-16-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty