Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dichloro-1-(3-isopropylphenyl)ethanol ≥97%

Supplier: AOB CHEM USA
Warning

55093-1G 55093-25G 55093-500MG 55093-5G
76891-744EA 233.4 USD InStock
76891-744 76891-746 76891-748 76891-750
2,2-Dichloro-1-(3-isopropylphenyl)ethanol ≥97%
2,2-Dichloro-1-(3-isopropylphenyl)ethanol


Formula: C₁₁H₁₄Cl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD34530354
CAS Number: 2734776-91-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty